Learn More
RPS6 Mouse anti-Human, Unconjugated, Clone: 1C3E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66886-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The RPS6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPS6. This antibody recognizes human, mouse, rat antigen. The RPS6 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
RPS6 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Mouse, Rat, Human |
Flow Cytometry, Immunofluorescence, Western Blot | |
1C3E10 | |
Unconjugated | |
20 μL | |
6194 | |
IgG2a |