missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS6KA3 (RSK2) Recombinant Human Protein
RPS6KA3 (RSK2) is a serine/threonine kinase.
Brand: Invitrogen™ PR5326A
Additional Details : Weight : 0.01000kg
Description
RPS6KA3 (RSK2) modulates many cellular processes including transcription, adhesion, growth, survival, and proliferation.
Specifications
NP_004577 | |
50mM Tris pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-10, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
RPS6KA3 (RSK2) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
86.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Baculovirus |