missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RPS6KA4 (MSK2), GST Tag Recombinant Protein
Human RPS6KA4 (MSK2), GST Tag Recombinant Protein
5380.00 SEK
Specifications
Accession Number | NP_003933 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 113.1 kDa |
Gene Alias | RPS6KA4 (MSK2) |
KinaseFamily | RSK (RPS6K) Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11543527
|
Invitrogen
PV3782 |
10μg |
5380.00 SEK
Each |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RPS6KA4 (MSK2) is a serine⁄threonine protein kinase that is essential for the regulation of various cellular processes. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos.Specifications
NP_003933 | |
113.1 kDa | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice |
Kinase Assay | |
RPS6KA4 (MSK2) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: AGC Group | |
Baculovirus |