missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS6KA6 (RSK4) Recombinant Human Protein
RPS6KA6 (RSK4) is a member of the RSK kinases family.
18240.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_055311 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 111.3kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15408663
|
Invitrogen™
PR8170A |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15408733
|
Invitrogen™
PV4558 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RSK family members are activated by ERKs and mitogens and are unusual in that they contain 2 non-identical kinase domains.
Specifications
NP_055311 | |
111.3kDa | |
RUO - research use only | |
RPS6KA6 (RSK4) | |
Human | |
GST Tag | |
Frozen |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Baculovirus |