Learn More
RPS6KA6 (RSK4) Recombinant Human Protein
RPS6KA6 (RSK4) Recombinant Human Protein
Brand:  Invitrogen PV4557
Additional Details : Weight : 0.50000kg
Product Code. 10516773
  • 4585.00 SEK / 10µg
Estimated Shipment: 11-02-2022
For Research Use Only. All usage must comply with product instructions.
Description
Description
RPS6KA6 (RSK4) is a member of the RSK kinases family. RSK family members are activated by ERKs and mitogens and are unusual in that they contain 2 non-identical kinase domains.
Specifications
Specifications
NP_055311
Kinase and inhibitors
Kinase assay
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
RPS6KA6 (RSK4) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
RPS6KA6 (RSK4)
RPS6KA6 (RSK4) was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL PAKtide peptide substrate (CRRKSLVGpTPYWMAPE) and trace [32P]-γ-ATP for 10 minutes at 30°C
RPS6KA6 (RSK4) Recombinant Human Protein
Human
Baculovirus
Serine/threonine kinase
0.44mg/mL
111.3kDa
90% by SDS-PAGE
–80°C
1 Each
RPS6KA6 (RSK4) Recombinant Human Protein
RSK (RPS6K)
1016nmoles of phosphate transferred to PAKtide peptide substrate (CRRKSLVGpTPYWMAPE) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.7μg/mL
GST Tag