Learn More
Invitrogen™ RPS6KB2 (p70S6K Beta) Recombinant Human Protein
RPS6KB2 (p70S6K beta) is a serine/threonine kinase with 2 non-identical kinase catalytic domains.
Brand: Invitrogen™ PR7468B
Additional Details : Weight : 0.01000kg
Description
RPS6KB2 (p70S6K beta) phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
Specifications
NP_003943 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
RPS6KB2 (p70S6K Beta) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
82.2kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Baculovirus |