missing translation for 'onlineSavingsMsg'
Learn More

RPS9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18215-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828274

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S4P family of ribosomal proteins. It is located in the cytoplasm. Variable expression of this gene in colorectal cancers compared to adjacent normal tissues has been observed, although no correlation between the level of expression and the severity of the disease has been found. As is typical for genes encoding ribosomal proteins, multiple processed pseudogenes derived from this gene are dispersed through the genome.
Specifications

Specifications

RPS9
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P46781, Q6ZWN5
40S ribosomal protein S9, ribosomal protein S9, RPS9
Rabbit
IgG
150 μL
-20°C
Primary
6203, 76846
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RPS9
Liquid
RPS9
RPS9 Fusion Protein Ag12945
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers