missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RPSA (Human) Recombinant Protein

Human RPSA full-length ORF ( AAH13827, 1 a.a. - 295 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00003921-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16177311

  • 3343.96 SEK / 10µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MSGALDVLQMKEEDVLKFLAAGTHLGGTNLDFQMEQYIYKRKSDGIYIINLKRTWEKLLLAARAIVAIENPADVSVISSRNTGQRAVLKFAAATGATPIAGRFTPGTFTNQIQAAFREPRLLVVTDPRAGHQPLTEASYVNLPTIALCNTDSPLRYVDIAIPCNNKGAHSVGLMWWMLAREVLRMRGTISREHPWEVMPDLYFYRDPEEIEKEEQAAAEKAVTKEEFQGEWTAPAPEFTATQPEVADWSEGVQVPSVPIQQFPTEDWSAQPATEDWSAAPTAQATEWVGATTDWS
Specifications

Specifications

AAH13827
ribosomal protein SA
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPSA
GST
3921
Wheat germ expression system
10 μg
37LRP, 67LR, LAMBR, LAMR1, LRP, p40
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates