Läs mer
(RS)-2-Ethylamino-N-(2-methylphenyl)propanamide Hydrochloride, Mikromol™
Produktkod. 30051063
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30051063 50 mg
1 options
Produktkod. 30051063 Leverantör Mikromol Leverantörsnummer MM0563.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(RS)-2-Ethylamino-N-(2-methylphenyl)propanamide Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (RS)-2-Ethylamino-N-(2-methylphenyl)propanamide Hydrochloride
CAS 35891-75-9
Analyt- eller komponentnamn (RS)-2-Ethylamino-N-(2-methylphenyl)propanamide Hydrochloride
Frakt skick Room Temperature
Molekylformel C12 H18 N2 O . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Prilocaine Hydrochloride Imp. C (EP), Prilocaine Imp. C (EP), (RS)-2-Ethylamino-N-(2-methylphenyl)propanamide Hydrochloride, Prilocaine Hydrochloride Imp. C (EP) as Hydrochloride, Prilocaine Hydrochloride Impurity C as Hydrochloride, Prilocaine Impurity C as Hydrochloride
InChI formel InChI=1S/C12H18N2O.ClH/c1-4-13-10(3)12(15)14-11-8-6-5-7-9(11)2;/h5-8,10,13H,4H2,1-3H3,(H,14,15);1H
LEDER Cl.CCNC(C)C(=O)Nc1ccccc1C
IUPAC-namn 2-(ethylamino)-N-(2-methylphenyl)propanamide;hydrochloride
Molekylvikt (g/mol) 242.75
Kvantitet 50 mg
Formel vikt 242.1186 g/mol