Läs mer
(RS)-3,5-DHPG, TRC
Produktkod. 30384432
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30384432 50 mg
30378768 10 mg
30380478 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30384432 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45002850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 146255-66-5
Molekylformel C8H9NO4
Molekylvikt (g/mol) 183.16
Synonym alpha-Amino-3,5-dihydroxybenzeneacetic Acid, (RS)-3,5-Dihydroxyphenylglycine, (±)-alpha-Amino-3,5-dihydroxybenzeneacetic Acid, 3,5-Dihydroxyphenylglycine, DHPG, DL-2-(3,5-Dihydroxyphenyl)glycine, (D/L)-3,5-Dihydroxyphenylglycine, 2-Amino-2-(3,5-dihydroxyphenyl)acetic Acid
LEDER NC(C(=O)O)c1cc(O)cc(O)c1
Specifikationer
Kemiskt namn eller material (RS)-3,5-DHPG
Kvantitet 50 mg
InChI formel InChI=1S/C8H9NO4/c9-7(8(12)13)4-1-5(10)3-6(11)2-4/h1-3,7,10-11H,9H2,(H,12,13)
Formel vikt 183.0532