Läs mer
RS 504393, MedChemExpress
Produktkod. 16439087
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16439087 5 mg -
16413476 25 mg 25mg
16406508 10 mg 10mg
16492597 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16439087 Leverantör Medchem Express Leverantörsnummer HY15418
MedChemExpress RS 504393 is a selective CCR2 chemokine receptor antagonist (IC50 values are 89 nM and > 100 μM for inhibition of human recombinant CCR2 and CCR1 receptors respectively).
RS 504393 is a selective CCR2 chemokine receptor antagonist (IC50 values are 89 nM and > 100 μM for inhibition of human recombinant CCR2 and CCR1 receptors respectively).
Specifikationer
Kemiskt namn eller material RS 504393
CAS 300816-15-3
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C25H27N3O3
Kvantitet 5 mg
Löslighetsinformation DMSO : 12.5 mg/mL (29.94 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
LEDER O=C1NC2=CC=C(C)C=C2C3(CCN(CCC4=C(C)OC(C5=CC=CC=C5)=N4)CC3)O1
Molekylvikt (g/mol) 417.5
Formel vikt 417.5
Procent renhet 98.06%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation