Läs mer
Tocris Bioscience™ (RS)-AMPA hydrobromide
GREENER_CHOICE
Produktkod. 18075352
Change view
Kvantitet:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18075352 10 mg
18094812 1 mg
18084272 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18075352 Leverantör Tocris Bioscience™ Leverantörsnummer 1074/10
Selective AMPA agonist; more water soluble form of (RS)-AMPA (Cat. No. 0169)
More water soluble hydrobromide salt of (RS)-AMPA (Cat. No. 0169).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 171259-81-7
Molekylformel C7H11BrN2O4
Specifikationer
Kemiskt namn eller material (RS)-AMPA hydrobromide
Kvantitet 10 mg
Formel vikt 267.08
Renhet >99%
Molekylvikt (g/mol) 267.079