missing translation for 'onlineSavingsMsg'
Läs mer

(RS)-N-Phenyl-2-(propylamino)propanamide, Mikromol™

Produktkod. 30049562
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049562 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30049562 Leverantör Mikromol Leverantörsnummer MM0563.06

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(RS)-N-Phenyl-2-(propylamino)propanamide
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material (RS)-N-Phenyl-2-(propylamino)propanamide
CAS 42459-45-0
Analyt- eller komponentnamn (RS)-N-Phenyl-2-(propylamino)propanamide
Frakt skick Room Temperature
Molekylformel C12 H18 N2 O
Certifikat och efterlevnad ISO 17025
Synonym (RS)-N-Phenyl-2-(propylamino)propanamide, Prilocaine Hydrochloride Impurity F, Prilocaine Impurity F
InChI formel InChI=1S/C12H18N2O/c1-3-9-13-10(2)12(15)14-11-7-5-4-6-8-11/h4-8,10,13H,3,9H2,1-2H3,(H,14,15)
LEDER CCCNC(C)C(=O)Nc1ccccc1
IUPAC-namn N-phenyl-2-(propylamino)propanamide
Molekylvikt (g/mol) 206.28
Kvantitet 100 mg
Formel vikt 206.1419 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.