Läs mer
Eurofins DiscoverX Rse Protein, Active
Beskrivning
Rse (TYRO3 protein tyrosine kinase), also known as Brt, BYK, Dtk, Etk3, Sky, Tif, or even sea-related receptor tyrosine kinase, is a receptor tyrosine kinase with homology to Axl. Rse is widely expressed in the adult and present in variable amounts in neural, lymphoid, vascular, and reproductive tissues. It plays a role in PI-3 kinase signaling and clearance of apoptotic cells. Gas6 acts directly on mature osteoclasts through activation of Tyro 3 and p42/p44 MAPK, possibly contributing to the bone loss by estrogen deficiency.
GenBank D17517
Specifikationer
Specifikationer
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekylvikt (g/mol) | 50.4 kDa |
| Kvantitet | 10 μg |
| Art | Human |
| Rekombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Uttryckssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Truncated |
| Renhet eller kvalitetsklass | ≥65% |
| Protein | Rse, active |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.