missing translation for 'onlineSavingsMsg'
Learn More

RSPO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17193-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877044

  • 4721.67 SEK / 150µL
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RSPO3 is a member of the R-spondin family and plays a role in the regulation of Wnt (wingless-type MMTV integration site family)/beta-catenin and Wnt/planar cell polarity (PCP) signaling pathways, which are involved in development, cell growth and disease pathogenesis. RSPO3 has been shown to be involved in regulating the balance between angioblast and blood cell specification during embryonic vasculogenesis and angiogenesis and may be involved in tumor development.
Specifications

Specifications

RSPO3
Polyclonal
Unconjugated
RSPO3
CRISTIN1, hPWTSR, hRspo3, Protein with TSP type 1 repeat, PWTSR, R spondin 3, Roof plate specific spondin 3, RSPO3, THSD2
Rabbit
Antigen Affinity Chromatography
RUO
498997, 72780, 84870
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q2TJ95, Q9BXY4
Rspo3
RSPO3 Fusion Protein Ag9899
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers