missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSRC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23826-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The arginine/serine-rich coiled-coil 1 protein (RSRC1) was initially identified as a non-snRNP splicing factor that has a role in both the constitutive and alternative splicing of pre-mRNA molecules. In cellular nuclear extracts immunodepleted of RSRC1, the second step of pre-mRNA splicing is blocked; recombit RSRC1 is able to restore splicing activity. RSRC1 has also been reported to be associated with schizophrenia as well as a unique marker of bone marrow-derived stem cells that also express the cdc34 marker.Specifications
RSRC1 | |
Polyclonal | |
Unconjugated | |
RSRC1 | |
BM 011, RSRC1, SRrp53 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51319 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96IZ7 | |
RSRC1 | |
RSRC1 Fusion Protein Ag20604 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |