missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSU1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RSU1 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828133
|
Proteintech
11207-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818133
|
Proteintech
11207-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Component of the HBO1 complex which has a histone H4-specific acetyltransferase activity, a reduced activity toward histone H3 and is responsible for the bulk of histone H4 acetylation in vivo. Through chromatin acetylation it may regulate DNA replication and act as a coactivator of TP53-dependent transcription. Specifically represses AR-mediated transcription.Specifications
RSU1 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q01730, Q15404 | |
20163, 6251 | |
RSU1 Fusion Protein Ag1693 | |
Primary | |
-20°C | |
RSU1 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Ras suppressor protein 1, RSP 1, RSP1, Rsu 1, RSU1 | |
RSU1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |