Learn More
Thermo Scientific™ Accessories for RT Basic Series and RT Touch Series Magnetic Stirrers
Use these accessories to optimize performance of your Thermo Scientific™ RT Basic Series Magnetic Stirrers.
Brand:  Thermo Scientific™ 88880144
Additional Details : Weight : 0.28000kg
 View more versions of this product
Product Code. 15208904
  • 102.00 SEK / Each
Estimated Shipment
30-04-2021
Add to basket
Specifications
White
RT Basic Series and RT Touch Series Magnetic Stirrers
Plate Cover
Non-slip silicone plate cover for items 88880007 and 88880008 (White color, Silicone 120mm dia.), Available for RT Basic Series only
96-Well Plates|384-Well Plates