Läs mer
RTI-51 Hydrochloride, TRC
Produktkod. 30416832
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30416832 25 mg
1 options
Produktkod. 30416832 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR70100025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R,2S,3S,5S)-3-(4-Bromophenyl)-8-methyl-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester Hydrochloride
CAS 1391052-88-2
Molekylformel C16H21BrClNO2
Kvantitet 25 mg
Synonym [1R-(exo,exo)]-3-(4-Bromophenyl)-8-methyl-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester Hydrochloride, RTI 4229-51 Hydrochloride, RTI-51 Hydrochloride
InChI formel InChI=1S/C16H20BrNO2.ClH/c1-18-12-7-8-14(18)15(16(19)20-2)13(9-12)10-3-5-11(17)6-4-10;/h3-6,12-15H,7-9H2,1-2H3;1H/t12-,13+,14+,15-;/m0./s1
LEDER Cl.COC(=O)[C@@H]1[C@H]2CC[C@@H](C[C@@H]1c3ccc(Br)cc3)N2C
IUPAC-namn methyl (1S,3S,4S,5R)-3-(4-bromophenyl)-8-methyl-8-azabicyclo[3.2.1]octane-4-carboxylate;hydrochloride
Molekylvikt (g/mol) 374.7
Formel vikt 373.0444