Learn More
DWK Life Sciences Rubber Adapter, from EPDM, for Filter Crucible
The rubber sleeve is required to achieve gas tightness during vacuum filtration.
259.00 SEK - 364.00 SEK
Specifications
Quantity 10/Pk.
Type Adapter
View More Specs
Products 4
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
11810720
View Documents Promotion Details
DWK Life Sciences
292013109
49 mm
364.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
11800720
View Documents Promotion Details
DWK Life Sciences
292012601
41 mm
345.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
11890710
View Documents Promotion Details
DWK Life Sciences
292012104
33 mm
333.00 SEK
Pack of 10
Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 10-05-2024
to see stock.
Añadir a la cesta
11880710
missing translation for 'viewDocuments' Detalles de la promoción
DWK Life Sciences
292011408
26 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Descripción
Descripción
In order to be able to connect a filter and a filtering flask together, a rubber sleeve is required which protects both glass parts from damage on the one hand and connects them gas-tight on the other hand. This gas tightness is required to perform a vacuum filtration.
  • For tight closure in vacuum filtration applications
  • For filter crucibles
Especificaciones
Especificaciones
10/Pk.
Adapter
Videos
SDS
Documentos
Documentos
Certificados