Learn More
Lovibond™ Rubber Gasket for Oxygen Meter
Accessory for BD 600, BD 606 and Oxi700 BOD Oxygen Measurment Systems. Biochemical Oxygen Demand (BOD) measurement is used as a basis for the detection of biologically degradable organic matter in water.
Brand:  Lovibond™ 418636
Additional Details : Weight : 0.00300kg
Product Code. 10293342
  • 106.00 SEK / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
Accessory for BD 600, BD 606 and Oxi700 BOD Oxygen Measurment Systems. Biochemical Oxygen Demand (BOD) measurement is used as a basis for the detection of biologically degradable organic matter in water.
  • Rubber Gasket
  • For BD 600, BD 606 and Oxi700 BOD Oxygen Meters
Specifications
Specifications
Oxygen Measument
BD 600, BD 606 and Oxi700 BOD Oxygen Measurment Systems
Rubber Gasket for Oxygen Meter