Learn More
Saint-Gobain Rubber Piston Ring
Use these rubber piston rings for a variety of applications. Saint Gobain Performance Plastics™ Rubber Piston Rings are available in a full complement of solid and split joint configurations.
Brand:  Saint-Gobain 101056
Additional Details : Weight : 0.60000kg
Product Code. 11774608
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Sturdy rubber piston rings for flasks
  • For general lab applications
Specifications
Specifications
Black
For 50mL to 500mL flask
Piston ring
Rubber
40 mm
80/120 mm