Läs mer
Rubusoside, TRC
Produktkod. 30134729
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134729 50 mg
30004493 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134729 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR70154550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 64849-39-4
Molekylformel C32 H50 O13
Molekylvikt (g/mol) 642.73
Synonym Kaur-16-en-18-oic acid, 13-(β-D-glucopyranosyloxy)-, β-D-glucopyranosyl ester, (4α)-, 1H-2,10a-Ethanophenanthrene, kaur-16-en-18-oic acid deriv., Rubusoside
LEDER C[C@@]12CCC[C@](C)([C@H]1CC[C@]34CC(=C)[C@](CC[C@@H]23)(C4)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C(=O)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O
Specifikationer
Kemiskt namn eller material Rubusoside
Kvantitet 50 mg
InChI formel InChI=1S/C32H50O13/c1-15-11-31-9-5-18-29(2,7-4-8-30(18,3)28(41)44-26-24(39)22(37)20(35)16(12-33)42-26)19(31)6-10-32(15,14-31)45-27-25(40)23(38)21(36)17(13-34)43-27/h16-27,33-40H,1,4-14H2,2-3H3/t16-,17-,18+,19+,20-,21-,22+,23+,24-,25-,26+,27+,29-,30-,31-,32+/m1/s1
Formel vikt 642.3251