Läs mer
Rucaparib phosphate, MedChemExpress
Produktkod. 16490996
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
200 mg
Förpackningsstorlek:
100mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16490996 200 mg -
16484068 10 mg -
16445046 100 mg 100mg
16440427 50 mg -
16406777 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16490996 Leverantör Medchem Express Leverantörsnummer HY10617
MedChemExpress Rucaparib (AG014699) phosphate is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP1. Rucaparib phosphate is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib phosphate has the potential for castration-resistant prostate cancer (CRPC) research.
Rucaparib (AG014699) phosphate is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP1. Rucaparib phosphate is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib phosphate has the potential for castration-resistant prostate cancer (CRPC) research.
Specifikationer
Kemiskt namn eller material Rucaparib phosphate
CAS 459868-92-9
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C19H21FN3O5P
Kvantitet 200 mg
Synonym AG-014699 phosphatePF-01367338 phosphate
Löslighetsinformation DMSO : ≥ 33 mg/mL (78.32 mM) ∣H2O : 5 mg/mL (11.87 mM; ultrasonic and warming and heat to 60°C)
LEDER O=P(O)(O)O.FC1=CC2=C3C(CCNC2=O)=C(C4=CC=C(CNC)C=C4)NC3=C1
Molekylvikt (g/mol) 421.36
Formel vikt 421.36
Procent renhet 98.77%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death