Läs mer
Rucaparib, TRC
Produktkod. 30132451
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132451 50 mg
30133183 10 mg
30006972 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132451 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR70158050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 283173-50-2
Molekylformel C19H18FN3O
Molekylvikt (g/mol) 323.36
Synonym 8-Fluoro-1,3,4,5-tetrahydro-2-[4-[(methylamino)methyl]phenyl]-6H-pyrrolo[4,3,2-ef][2]benzazepin-6-one, 8-Fluoro-2-[4-[(methylamino)methyl]phenyl]-1,3,4,5-tetrahydro-6H-azepino[5,4,3-cd]indol-6-one, Rubraca, 8-Fluoro-1,3,4,5-tetrahydro-2-[4-[(methylamino)methyl]phenyl]-6H-azepino[5,4,3-cd]indol-6-one;
LEDER CNCc1ccc(cc1)c2[nH]c3cc(F)cc4C(=O)NCCc2c34
Specifikationer
Kemiskt namn eller material Rucaparib
Kvantitet 50 mg
InChI formel InChI=1S/C19H18FN3O/c1-21-10-11-2-4-12(5-3-11)18-14-6-7-22-19(24)15-8-13(20)9-16(23-18)17(14)15/h2-5,8-9,21,23H,6-7,10H2,1H3,(H,22,24)
Formel vikt 323.1434