Läs mer
Rufinamide-15N,d2, TRC
Product Code. 30420035
Change view
Click to view available options
Kvantitet:
1 mg
Product Code. Kvantitet
30420035 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30420035 Supplier Toronto Research Chemicals Supplier No. TRCR7015521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Rufinamide-15N,d2
CAS 1795037-48-7
Anteckningar om renhetsgrad HPLC
Molekylformel C10H6D2F2N315NO
Kvantitet 1 mg
Synonym 1-[(2,6-Difluorophenyl)methyl]-1H-1,2,3-triazole-4-carboxamide-15N,d2, CGP 33101-15N,d2, E 2080-15N,d2, Inovelon, RUF 331-15N,d2
InChI formel InChI=1S/C10H8F2N4O/c11-7-2-1-3-8(12)6(7)4-16-5-9(10(13)17)14-15-16/h1-3,5H,4H2,(H2,13,17)/i4D2,13+1
LEDER FC1=CC=CC(F)=C1C([2H])([2H])N2C=C(C([15NH2])=O)N=N2
IUPAC-namn 1-[dideuterio-(2,6-difluorophenyl)methyl]triazole-4-(^{15}N)carboxamide
Molekylvikt (g/mol) 241.2
Formel vikt 241.0762