Läs mer
Rufinamide-5-Carboxamide, TRC
Produktkod. 30425535
Change view
Kvantitet:
0.5 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30425535 2.5 mg 2.5mg
30410978 5 mg -
30418929 0.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR7014702.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1622904-99-7
Molekylformel C10H8F2N4O
Molekylvikt (g/mol) 238.19
Synonym Rufinamide Impurity 3
IUPAC-namn 3-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
LEDER NC(=O)c1cnnn1Cc2c(F)cccc2F
Specifikationer
Kemiskt namn eller material Rufinamide-5-Carboxamide
Kvantitet 2.5 mg
InChI formel InChI=1S/C10H8F2N4O/c11-7-2-1-3-8(12)6(7)5-16-9(10(13)17)4-14-15-16/h1-4H,5H2,(H2,13,17)
Formel vikt 238.0666