Läs mer
Rupintrivir, TRC
Produktkod. 30412827
Change view
Kvantitet:
0.25 mg
2.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412827 0.25 mg
30409389 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30412827 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR7017000.25MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 223537-30-2
Molekylformel C31H39FN4O7
Molekylvikt (g/mol) 598.66
Synonym (2E,4S)-4-[[(2R,5S)-2-[(4-Fluorophenyl)methyl]-6-methyl-5-[[(5-methyl-3-isoxazolyl)carbonyl]amino]-1,4-dioxoheptyl]amino]-5-[(3S)-2-oxo-3-pyrrolidinyl]-2-pentenoic Acid, AG 7088
IUPAC-namn ethyl (E,4S)-4-[[(2R,5S)-2-[(4-fluorophenyl)methyl]-6-methyl-5-[(5-methyl-1,2-oxazole-3-carbonyl)amino]-4-oxoheptanoyl]amino]-5-[(3S)-2-oxopyrrolidin-3-yl]pent-2-enoate
LEDER CCOC(=O)\C=C\[C@H](C[C@@H]1CCNC1=O)NC(=O)[C@@H](CC(=O)[C@@H](NC(=O)c2cc(C)on2)C(C)C)Cc3ccc(F)cc3
Specifikationer
Kemiskt namn eller material Rupintrivir
Kvantitet 0.25 mg
InChI formel InChI=1S/C31H39FN4O7/c1-5-42-27(38)11-10-24(16-21-12-13-33-29(21)39)34-30(40)22(15-20-6-8-23(32)9-7-20)17-26(37)28(18(2)3)35-31(41)25-14-19(4)43-36-25/h6-11,14,18,21-22,24,28H,5,12-13,15-17H2,1-4H3,(H,33,39)(H,34,40)(H,35,41)/b11-10+/t21-,22+,24+,28-/m0/s1
Formel vikt 598.2803