Learn More
Ruxolitinib Phosphate, TRC
Product Code. 17993345
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993345 10 mg 10mg
30135821 100 mg 100mg
17925045 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17993345 Supplier Toronto Research Chemicals Supplier No. TRCR70200510MG
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1092939-17-7
Molecular Formula C17 H18 N6 . H3 O4 P
Molecular Weight (g/mol) 404.36
Synonym 1H-Pyrazole-1-propanenitrile, β-cyclopentyl-4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-, (βR)-, phosphate (1:1), INC 424 phosphate, INCB 018424 phosphate, Jakafi, Ruxolitinib phosphate
IUPAC Name (3R)-3-cyclopentyl-3-[4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)pyrazol-1-yl]propanenitrile;phosphoric acid
SMILES OP(=O)(O)O.N#CC[C@H](C1CCCC1)n2cc(cn2)c3ncnc4[nH]ccc34
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C17H18N6.H3O4P/c18-7-5-15(12-3-1-2-4-12)23-10-13(9-22-23)16-14-6-8-19-17(14)21-11-20-16;1-5(2,3)4/h6,8-12,15H,1-5H2,(H,19,20,21);(H3,1,2,3,4)/t15-;/m1./s1
Formula Weight 404.1362
Percent Purity >95%
Chemical Name or Material Ruxolitinib Phosphate
Grade HPLC