Learn More
Invitrogen™ RXRA Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
Retinoid X receptor alpha, ligand binding domain (RXRA-LBD) is a 56.67 kDa recombinant human protein (amino acids 204-462) expressed as a GST fusion protein in baculovirus-infected insect cells.
Brand: Invitrogen™ PV4799
Additional Details : Weight : 0.00010kg
Description
RXRA-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
- Functionally confirmed in a LanthaScreen™ TR-FRET coactivator assay to ensure performance
- Storage buffer: 50mM Tris-HCl pH 8.0, 100mM KCl, 0.5mM EDTA, 5mM DTT, 20% glycerol
Specifications
Nuclear Receptor Assay | |
50 μg | |
RUO - research use only | |
Retinoid X Receptor alpha | |
Recombinant |
6256 | |
Store in ultra-cold freezer (-68 to -85°C). | |
Recombinant, Ligand Binding Domain | |
Human | |
Hormone Receptors |