Learn More
Invitrogen™ RXRB Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
Retinoid X receptor beta, ligand binding domain (RXRB-LBD) is a 54.5 kDa recombinant human protein (amino acids 295-533) expressed as a GST-tagged fusion protein in insect cells.
Brand: Invitrogen™ PV4390
Additional Details : Weight : 0.50000kg
Description
RXRB-LBD is expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator recruitment studies.
Specifications
Nuclear Receptor Assay | |
50 μg | |
RUO - research use only | |
Retinoid X Receptor Beta | |
Recombinant | |
Hormone Receptors |
6257 | |
Store in ultra-cold freezer (-68 to -85°C). | |
Recombinant, Ligand Binding Domain | |
Human | |
GST Tag |