Learn More
Invitrogen™ RXRB Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
Retinoid X receptor beta, ligand binding domain (RXRB-LBD) is a 54.5 kDa recombinant human protein (amino acids 295-533) expressed as a GST-tagged fusion protein in insect cells.
Brand:  Invitrogen™ PV4390
Additional Details : Weight : 0.50000kg
Product Code. 10577543
  • 17690.00 SEK / 50µg
Estimated Shipment: 15-04-2024
to see stock.
For Research Use Only. All usage must comply with product instructions.
Description
Description
RXRB-LBD is expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator recruitment studies.
Specifications
Specifications
Nuclear Receptor Assay
50 μg
RUO - research use only
Retinoid X Receptor Beta
Recombinant
Hormone Receptors
6257
Store in ultra-cold freezer (-68 to -85°C).
Recombinant, Ligand Binding Domain
Human
GST Tag