missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RYBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RYBP |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828393
|
Proteintech
11365-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818393
|
Proteintech
11365-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RYBP involves component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1-like complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A ′Lys-119′, rendering chromatin heritably changed in its expressibility. Component of a PRC1-like complex that mediates monoubiquitination of histone H2A ′Lys-119′ on the X chromosome and is required for normal silencing of one copy of the X chromosome in XX females. May stimulate ubiquitination of histone H2A ′Lys-119′ by recruiting the complex to target sites. Inhibits ubiquitination and subsequent degradation of TP53, and thereby plays a role in regulating transcription of TP53 target genes. May also regulate the ubiquitin-mediated proteasomal degradation of other proteins like FANK1 to regulate apoptosis. May be implicated in the regulation of the transcription as a repressor of the transcriptional activity of E4TF1. May bind to DNA. May play a role in the repression of tumor growth and metastasis in breast cancer by down-regulating SRRM3.Specifications
RYBP | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q8CCI5, Q8N488 | |
23429, 312603, 56353 | |
RYBP Fusion Protein Ag1908 | |
Primary | |
-20°C | |
RYBP |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AAP1, APAP 1, Apoptin associating protein 1, DED associated factor, DEDAF, RING1 and YY1 binding protein, RYBP, YEAF1 | |
Rybp | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |