missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-1-(2-Naphthyl)ethylamine, ChiPros 99+%, ee 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
(S)-(-)-1-(2-Naphthyl)ethylamine is used in asymmetric synthesis. It is also used in the preparation of thiourea compounds using chiral amine, which is used as a bifunctional catalyst for the Strecker and nitro-Michael reactions. Further, it acts as a chiral building block and chiral resolution reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(-)-1-(2-Naphthyl)ethylamine is used in asymmetric synthesis. It is also used in the preparation of thiourea compounds using chiral amine, which is used as a bifunctional catalyst for the Strecker and nitro-Michael reactions. Further, it acts as a chiral building block and chiral resolution reagent.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Sensitive to carbon dioxide.
(S)-(-)-1-(2-Naphthyl)ethylamine is used in asymmetric synthesis. It is also used in the preparation of thiourea compounds using chiral amine, which is used as a bifunctional catalyst for the Strecker and nitro-Michael reactions. Further, it acts as a chiral building block and chiral resolution reagent.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Sensitive to carbon dioxide.
Specifications
Melting Point | 48°C to 52°C |
Boiling Point | 90°C (0.15 mmHg) |
Quantity | 1 g |
UN Number | UN3259 |
Beilstein | 3195949 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Optical Rotation | −20° (c=1 in Methanol) |
Formula Weight | 171.24 |
Percent Purity | ≥99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'