Läs mer
(S)-1-(3-Bromophenyl)ethylamine, ChiPros™ 99%, ee 98+%
Produktkod. p-7029527
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11461640 1 g 1g
11471640 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11461640 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26998.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 139305-96-7 | C8H10BrN | 200.079 g/mol
(S)-1-(3-Bromophenyl)ethylamine is involved in the Ullmann coupling reaction with N-H containing heteroarenes using copper iodide as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-1-(3-Bromophenyl)ethylamine is involved in the Ullmann coupling reaction with N-H containing heteroarenes using copper iodide as a catalyst.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents, carbon monoxide and strong acids.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139305-96-7
Molekylformel C8H10BrN
Molekylvikt (g/mol) 200.079
MDL-nummer MFCD00671638
InChI-nyckel LIBZHYLTOAGURM-LURJTMIESA-N
Synonym s-1-3-bromophenyl ethanamine, s-1-3-bromophenyl ethylamine, 1s-1-3-bromophenyl ethanamine, 1s-1-3-bromophenyl ethan-1-amine, s-3-bromo-alpha-methylbenzylamine, 1s-1-3-bromophenyl ethylamine, s---3-bromo alpha-methylbenzylamine, s-m-bromo-, a-methylbenzylamine, s-3-bromo-
PubChem CID 9952938
IUPAC-namn (1S)-1-(3-bromophenyl)ethanamine
LEDER CC(C1=CC(=CC=C1)Br)N
Specifikationer
Kemiskt namn eller material (S)-1-(3-Bromophenyl)ethylamine
Densitet 1.33
Kokpunkt 96°C (4 mmHg)
Flampunkt >110°C (230°F)
Anteckningar om analysprocent ee ≥98%
Kvantitet 1 g
UN-nummer UN2735
Känslighet Air Sensitive
Löslighetsinformation Immiscible with water.
Optisk rotation −22° (Neat)
Formel vikt 200.08
Procent renhet 99%