missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1-(4-Chlorophenyl)ethylamine, 95+%, (sum of isomers), 95% e.e., Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Density | 1.1150g/mL |
Boiling Point | 232.0°C |
Quantity | 1 g |
Packaging | Glass bottle |
Refractive Index | 1.5410 to 1.5430 |
Assay Percent Range | 95% min. (GC) |
Specific Gravity | 1.115 |
Linear Formula | ClC6H4CH(CH3)NH2 |
Solubility Information | Solubility in water: moderately soluble |
Formula Weight | 155.63 |
Show More |
Safety and Handling
Product Identifier
- (S)-1-(4-Chlorophenyl)ethylamine
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'