Learn More
(S)-1,4-Diazabicyclo[4.3.0]nonane, 98%, Thermo Scientific™
Catalog Number p-7077640
Click to view available options
Quantity:
250 mg
1 g
93643-24-4
C7H14N2
126.20
MFCD03787926
FTTATHOUSOIFOQ-SSDOTTSWSA-N
s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane
781249
(8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
C1C[C@@H]2CNCCN2C1
93643-24-4
C7H14N2
126.20
MFCD03787926
FTTATHOUSOIFOQ-SSDOTTSWSA-N
s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane
781249
(8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
C1C[C@@H]2CNCCN2C1
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 250 mg
Specific Rotation Condition + 5.88 (20.00°C c=1, CHCl3)
Specific Rotation + 5.88
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material (S)-1,4-Diazabicyclo[4.3.0]nonane
compliance-icons
Product Identifier
  • (S)-1,4-Diazabicyclo¢4.3.0!nonane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.