Läs mer
(S)-1-(p-Bromoacetamidobenzyl)ethylenediaminetetraacetic Acid (~80%), TRC
Produktkod. 30340732
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30340732 5 mg
30344003 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30340732 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6790005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-1-(p-Bromoacetamidobenzyl)ethylenediaminetetraacetic acid
CAS 81677-64-7
Molekylformel C19 H24 Br N3 O9
Kvantitet 5 mg
InChI formel InChI=1S/C19H24BrN3O9/c20-6-15(24)21-13-3-1-12(2-4-13)5-14(23(10-18(29)30)11-19(31)32)7-22(8-16(25)26)9-17(27)28/h1-4,14H,5-11H2,(H,21,24)(H,25,26)(H,27,28)(H,29,30)(H,31,32)/t14-/m0/s1
LEDER OC(=O)CN(C[C@H](Cc1ccc(NC(=O)CBr)cc1)N(CC(=O)O)CC(=O)O)CC(=O)O
IUPAC-namn 2-[[(2S)-2-[bis(carboxymethyl)amino]-3-[4-[(2-bromoacetyl)amino]phenyl]propyl]-(carboxymethyl)amino]acetic acid
Molekylvikt (g/mol) 518.31
Formel vikt 517.0696