Learn More
(S)-(-)-1-Phenylethyl isocyanate, 98%, Thermo Scientific Chemicals
Catalog Number p-7045557
Click to view available options
Quantity:
1 g
5 g
14649-03-7
C9H9NO
147.177
MFCD00002036
JJSCUXAFAJEQGB-QMMMGPOBSA-N
s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
2724173
[(1S)-1-isocyanatoethyl]benzene
CC(C1=CC=CC=C1)N=C=O
14649-03-7
C9H9NO
147.177
MFCD00002036
JJSCUXAFAJEQGB-QMMMGPOBSA-N
s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
2724173
[(1S)-1-isocyanatoethyl]benzene
CC(C1=CC=CC=C1)N=C=O
(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY
Density 1.045
Boiling Point 55°C to 56°C (2.5 mmHg)
Flash Point 65°C (149°F)
Odor Pungent
Refractive Index 1.5145
Quantity 1 g
UN Number UN2206
Beilstein 4230972
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Hydrolyzes in water.
Optical Rotation −10.5° (Neat)
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material (S)-(-)-1-Phenylethyl isocyanate