Läs mer
(S)-1-Trichloromethyl-1,2,3,4-Tetrahydro-Beta-carboline Hydrochloride, TRC
Produktkod. 30456702
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456702 10 mg
30457105 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30456702 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77424010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 773057-50-4
Molekylformel C12H11Cl3N2 . HCl
Molekylvikt (g/mol) 326
Synonym (S)-2,3,4,9-Tetrahydro-1-(trichloromethyl)-1H-pyrido[3,4-b]indole Hydrochloride, (R)-TaClo Hydrochloride
IUPAC-namn (1S)-1-(trichloromethyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole;hydrochloride
LEDER Cl.ClC(Cl)(Cl)[C@H]1NCCc2c1[nH]c3ccccc23
Specifikationer
Kemiskt namn eller material (S)-1-Trichloromethyl-1,2,3,4-Tetrahydro-beta-carboline Hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C12H11Cl3N2.ClH/c13-12(14,15)11-10-8(5-6-16-11)7-3-1-2-4-9(7)17-10;/h1-4,11,16-17H,5-6H2;1H/t11-;/m0./s1
Formel vikt 323.9755