Läs mer
(S)-[2-[[2-(2-Chlorobenzoyl)-4-nitrophenyl]amino]-1-methyl-2-oxoethyl]-carbamic Acid Phenylmethyl Ester, TRC
Produktkod. 30349979
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30349979 25 mg
1 options
Produktkod. 30349979 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36454325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-(2-((2-(2-Chlorobenzoyl)-4-nitrophenyl)amino)-1-methyl-2-oxoethyl)-carbamic Acid Phenylmethyl Ester
CAS 58662-82-1
Molekylformel C24H20ClN3O6
Kvantitet 25 mg
Synonym (S)-Benzyl (1-((2-(2-chlorobenzoyl)-4-nitrophenyl)amino)-1-oxopropan-2-yl)carbamate
InChI formel InChI=1S/C24H20ClN3O6/c1-15(26-24(31)34-14-16-7-3-2-4-8-16)23(30)27-21-12-11-17(28(32)33)13-19(21)22(29)18-9-5-6-10-20(18)25/h2-13,15H,14H2,1H3,(H,26,31)(H,27,30)/t15-/m0/s1
LEDER C[C@H](NC(=O)OCc1ccccc1)C(=O)Nc2ccc(cc2C(=O)c3ccccc3Cl)[N+](=O)[O-]
IUPAC-namn benzyl N-[(2S)-1-[2-(2-chlorobenzoyl)-4-nitroanilino]-1-oxopropan-2-yl]carbamate
Molekylvikt (g/mol) 481.89
Formel vikt 481.1041