Läs mer
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate, TRC
Produktkod. 30372461
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30372461 1 g
1 options
Produktkod. 30372461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4695551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate
CAS 23735-43-5
Molekylformel C13H18O5S
Kvantitet 1 g
Synonym 2,2-Dimethyl-1,3-dioxolane-4-methanol-4-methylbenzenesulfonate, 4-Methylbenzenesulfonic Acid (S)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Ester, Toluene-4-sulfonic Acid (S)-2,2-Dimethyl-[1,3]dioxolan-4-ylmethyl Ester, p-Toluenesulfonic Acid ((4S)-2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Ester
InChI formel InChI=1S/C13H18O5S/c1-10-4-6-12(7-5-10)19(14,15)17-9-11-8-16-13(2,3)18-11/h4-7,11H,8-9H2,1-3H3/t11-/m0/s1
LEDER Cc1ccc(cc1)S(=O)(=O)OC[C@@H]2COC(C)(C)O2
Molekylvikt (g/mol) 286.34