Learn More
(S)-2-acetamido-N-((S)-1-amino-4-methyl-1-oxopentan-2-yl)-4-methyl-N-((S)-1-oxohexan-2-yl)pentanamide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 110044-82-1 |
| Molecular Formula | C20H37N3O4 |
| MDL Number | MFCD00065505 |
| Quantity | 250 mg |
| SMILES | O=C(N[C@H](C=O)CCCC)[C@@H](NC([C@@H](NC(C)=O)CC(C)C)=O)CC(C)C |
| Molecular Weight (g/mol) | 383.53 |
| Percent Purity | 98% |
| Chemical Name or Material | (S)-2-acetamido-N-((S)-1-amino-4-methyl-1-oxopentan-2-yl)-4-methyl-N-((S)-1-oxohexan-2-yl)pentanamide |