Läs mer
(S)-2-Amino-4-methyl-1-((S)-2-methyloxiran-2-yl)pentan-1-one Trifluoroacetate Salt, TRC
Produktkod. 30312920
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30312920 50 mg
1 options
Produktkod. 30312920 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60510550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-2-Amino-4-methyl-1-((S)-2-methyloxiran-2-yl)pentan-1-one Trifluoroacetate Salt
CAS 1528769-14-3
Molekylformel C11H18F3NO4
Kvantitet 50 mg
Synonym 1-Pentanone, 2-amino-4-methyl-1-[(2S)-2-methyl-2-oxiranyl]-, (2S)-, 2,2,2-trifluoroacetate (1:1), (S)-2-Amino-4-methyl-1-((S)-2-methyloxiran-2-yl)pentan-1-one trifluoroacetate salt, (2S)-2-Amino-4-methyl-1-[(2S)-2-methyloxiran-2-yl]pentan-1-one trifluoroacetate
InChI formel InChI=1S/C9H17NO2.C2HF3O2/c1-6(2)4-7(10)8(11)9(3)5-12-9;3-2(4,5)1(6)7/h6-7H,4-5,10H2,1-3H3;(H,6,7)/t7-,9-;/m0./s1
LEDER CC(C)C[C@H](N)C(=O)[C@]1(C)CO1.OC(=O)C(F)(F)F
IUPAC-namn (2S)-2-amino-4-methyl-1-[(2S)-2-methyloxiran-2-yl]pentan-1-one;2,2,2-trifluoroacetic acid
Molekylvikt (g/mol) 285.26