Läs mer
(S)-2-Azido Isovaleric Acid Cyclohexylammonium Salt, TRC
Produktkod. 30333940
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30333940 1 g
30329149 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30333940 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA8547001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-2-Azido Isovaleric Acid Cyclohexylammonium Salt
CAS 1217462-63-9
Molekylformel C5H9N3O2
Kvantitet 1 g
Synonym (2S)-2-Azido-3-methylbutanoic Acid Cyclohexylammonium Salt, (S)-2-Azido-3-methylbutyric Acid Cyclohexylammonium Salt
InChI formel InChI=1S/C6H13N.C5H9N3O2/c7-6-4-2-1-3-5-6;1-3(2)4(5(9)10)7-8-6/h6H,1-5,7H2;3-4H,1-2H3,(H,9,10)/t;4-/m.0/s1
LEDER CC(C)[C@H](N=[N+]=[N-])C(=O)O.NC1CCCCC1
IUPAC-namn (2S)-2-azido-3-methylbutanoic acid;cyclohexanamine
Molekylvikt (g/mol) 143.14
Formel vikt 242.1743