Läs mer
(S)-2-Hydroxysuccinic Acid Methyl Ester, TRC
Produktkod. 30405605
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30405605 250 mg
1 options
Produktkod. 30405605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH953570250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-2-Hydroxysuccinic Acid Methyl Ester
CAS 66212-45-1
Molekylformel C5 H8 O5
Kvantitet 250 mg
Synonym Butanedioic acid, 2-hydroxy-, 1-methyl ester, (2S)-, Butanedioic acid, hydroxy-, 1-methyl ester, (2S)- (9CI), Butanedioic acid, hydroxy-, 1-methyl ester, (S)-, 1-Methyl (2S)-2-hydroxybutanedioate, (2S)-2-Hydroxy-butanedioic acid 1-mono methyl ester, (3S)-3-Hydroxy-4-methoxy-4-oxobutanoic acid, (S)-2-Hydroxysuccinic acid methyl ester, Methyl (S)-2-hydroxysuccinate
InChI formel InChI=1S/C5H8O5/c1-10-5(9)3(6)2-4(7)8/h3,6H,2H2,1H3,(H,7,8)/t3-/m0/s1
LEDER COC(=O)[C@@H](O)CC(=O)O
IUPAC-namn (3S)-3-hydroxy-4-methoxy-4-oxobutanoic acid
Molekylvikt (g/mol) 148.11
Formel vikt 148.0372