Läs mer
(S)-2-Methyl-1,4,5,6-tetrahydropyrimidine-4-carboxylic Acid, TRC
Produktkod. 30472526
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472526 500 mg
30465152 100 mg
30464684 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30472526 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM338018500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96702-03-3
Molekylformel C6H10N2O2
Molekylvikt (g/mol) 142.16
Synonym (4S)-3,4,5,6-Tetrahydro-2-methyl-4-pyrimidinecarboxylic Acid, (4S)-2-Methyl-1,4,5,6-tetrahydropyrimidine-4-carboxylic Acid, (4S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic Acid, (S)-2-methyl-1,4,5,6-tetrahydropyrimidine-4-carboxylic Acid, Ectoin, Ectoine, NSC 614616, Pyrostatin B, Pyrostatine B, RonaCare
IUPAC-namn (6S)-2-methyl-1,4,5,6-tetrahydropyrimidine-6-carboxylic acid
LEDER CC1=NCC[C@H](N1)C(=O)O
Specifications
Kemiskt namn eller material (S)-2-Methyl-1,4,5,6-tetrahydropyrimidine-4-carboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C6H10N2O2/c1-4-7-3-2-5(8-4)6(9)10/h5H,2-3H2,1H3,(H,7,8)(H,9,10)/t5-/m0/s1
Formel vikt 142.0742
Procent renhet >95%