Läs mer
(S)-2-Phenyl-1-propanamine, TRC
Produktkod. 30425708
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30425708 50 mg
1 options
Produktkod. 30425708 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP39917050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-2-Phenyl-1-propanamine
CAS 17596-79-1
Molekylformel C9H13N
Kvantitet 50 mg
Synonym (S)-2-Phenylpropylamine, (S)-beta-Methylphenethylamine, S-(-)-2-Phenylpropylamine, (betaS)-beta-Methylbenzeneethanamine, ((2S)-2-Phenylpropyl)amine, (-)-2-Phenylpropylamine, (-)-beta-Methylphenethylamine, (2S)-2-Phenylpropan-1-amine, (2S)-Phenylpropan-1-amine
InChI formel InChI=1S/C9H13N/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3/t8-/m1/s1
LEDER C[C@H](CN)C1=CC=CC=C1
IUPAC-namn (2S)-2-phenylpropan-1-amine
Molekylvikt (g/mol) 135.21
Formel vikt 135.1048