missing translation for 'onlineSavingsMsg'
Läs mer

(S)-3-(2-Bromo-4-methoxyphenyl)-2-(4-methoxyphenyl)propan-1-ol, TRC

Produktkod. 30344906
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30344906 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30344906 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB684760500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material (S)-3-(2-Bromo-4-methoxyphenyl)-2-(4-methoxyphenyl)propan-1-
CAS 917379-11-4
Molekylformel C17 H19 Br O3
Kvantitet 500 mg
InChI formel InChI=1S/C17H19BrO3/c1-20-15-6-3-12(4-7-15)14(11-19)9-13-5-8-16(21-2)10-17(13)18/h3-8,10,14,19H,9,11H2,1-2H3/t14-/m1/s1
LEDER COc1ccc(cc1)[C@@H](CO)Cc2ccc(OC)cc2Br
IUPAC-namn (2S)-3-(2-bromo-4-methoxyphenyl)-2-(4-methoxyphenyl)propan-1-ol
Molekylvikt (g/mol) 351.24
Formel vikt 350.0518
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.