Läs mer
(S)-3,5-DHPG, Tocris Bioscience™
Selective group I mGlu agonist; active enantiomer of 3,5-DHPG (Cat. No. 0342)
Brand: Tocris Bioscience 0805/5
Beskrivning
Selective group I mGlu receptor agonist.Available as part of the Group I mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . Racemate also available.
Specifikationer
| (S)-3, 5-DHPG | |
| C8H9NO4 | |
| s-3,5-dihydroxyphenylglycine, s-3,5-dhpg, l-3,5-dihydroxyphenylglycine, s-dhpg, unii-cf5g2g268a, chembl39221, 2s-2-amino-2-3,5-dihydroxyphenyl acetic acid, benzeneacetic acid, alpha-amino-3,5-dihydroxy-, alphas, tocris-0342, tocris-0805 | |
| NC(C(O)=O)C1=CC(O)=CC(O)=C1 | |
| 183.16 | |
| 443586 | |
| 183.16 |
| 162870-29-3 | |
| MFCD11044457 | |
| HOOWCUZPEFNHDT-UHFFFAOYNA-N | |
| 2-amino-2-(3,5-dihydroxyphenyl)acetic acid | |
| 5mg | |
| CHEBI:29474 | |
| >98% |