missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7055037
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
188970-92-5
C22H22N2O6
410.426
MFCD00273468
MPVGCCAXXFLGIU-IBGZPJMESA-N
fmoc-dap alloc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-allyloxy carbonyl amino propanoic acid, fmoc-dap aloc-oh, fmoc-3-allyloxy carbonyl amino-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-prop-2-en-1-yloxy carbonyl amino propanoic acid, ambotzfaa1366, fmoc-3-allyloxycarbonylamino-l-ala-oh, fmoc-dap alloc-oh hplc, n-, a-fmoc-n-, a-allyloxycarbonyl-l-2,3-diaminopropion, na-fmoc-nb-allyloxycarbonyl-l-2,3-diaminopropionic acid
2756107
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(prop-2-enoxycarbonylamino)propanoic acid
C=CCOC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
This item is not returnable. View return policy
188970-92-5
C22H22N2O6
410.426
MFCD00273468
MPVGCCAXXFLGIU-IBGZPJMESA-N
fmoc-dap alloc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-allyloxy carbonyl amino propanoic acid, fmoc-dap aloc-oh, fmoc-3-allyloxy carbonyl amino-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-prop-2-en-1-yloxy carbonyl amino propanoic acid, ambotzfaa1366, fmoc-3-allyloxycarbonylamino-l-ala-oh, fmoc-dap alloc-oh hplc, n-, a-fmoc-n-, a-allyloxycarbonyl-l-2,3-diaminopropion, na-fmoc-nb-allyloxycarbonyl-l-2,3-diaminopropionic acid
2756107
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(prop-2-enoxycarbonylamino)propanoic acid
C=CCOC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 9161065
Formula Weight 410.42
Percent Purity 95%
Physical Form Solid
Chemical Name or Material (S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'